Learn More
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
£51.36 - £223.11
Chemical Identifiers
CAS | 87-69-4 |
---|---|
Molecular Formula | C4H6O6 |
Molecular Weight (g/mol) | 150.09 |
MDL Number | MFCD00064207 |
InChI Key | FEWJPZIEWOKRBE-UHFFFAOYNA-N |
Synonym | l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid |
PubChem CID | 444305 |
ChEBI | CHEBI:15671 |
IUPAC Name | 2,3-dihydroxybutanedioic acid |
SMILES | OC(C(O)C(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10455091
|
Thermo Scientific Acros
420005000 |
500 g | Plastic bottle |
£51.36
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10689223
|
Thermo Scientific Acros
420000025 |
2.5 kg | Plastic bottle |
£223.11
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87-69-4 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
444305 | |
2,3-dihydroxybutanedioic acid |
C4H6O6 | |
MFCD00064207 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
CHEBI:15671 | |
OC(C(O)C(O)=O)C(O)=O |
Specifications
87-69-4 | |
C4H6O6 | |
MFCD00064207 | |
16,304 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
OC(C(O)C(O)=O)C(O)=O | |
150.09 | |
CHEBI:15671 | |
≥99.0% | |
Authentic | |
0.02% max. |
White | |
HO2CCH(OH)CH(OH)CO2H | |
03, 481 | |
15, 9205 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
2,3-dihydroxybutanedioic acid | |
444305 | |
150.09 | |
ACS Reagent | |
Crystalline | |
L(+)-Tartaric acid, 99+% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 201-766-0
missing translation for 'rtecsNumber' : WW7875000
missing translation for 'tsca' : TSCA
- L(+)-Tartaric acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : (+)-Tartaric acid
RUO – Research Use Only