missing translation for 'onlineSavingsMsg'
Learn More
L(+)-Tartaric acid, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals Product Code.: 15730869

L(+)-Tartaric acid, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Product Code. 15730869
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 15730869

Brand: Thermo Scientific Acros 388280010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87-69-4
Melting Point 168.0°C to 172.0°C
Molecular Formula C4H6O6
Linear Formula HO2CCH(OH)CH(OH)CO2H
MDL Number MFCD00064207
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
Solubility Information Solubility in water: 1390g/L (20°C). Other solubilities: soluble in methanol, ethanol, propanol and, glycerol, 4g/L ether, insoluble in chloroform
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES OC(C(O)C(O)=O)C(O)=O
IUPAC Name 2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
PubChem CID 444305
ChEBI CHEBI:15671
Formula Weight 150.09
Quantity 1 kg
Flash Point 210°C
Loss on Drying 0.2% max. (1 g, 105°C)
Chemical Name or Material L(+)-Tartaric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465915 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog Number388280010
Lot NumberA0450757 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number388280010
Lot NumberA0446139 Certificate TypeCertificate of Analysis Date21/09/2022 Catalog Number388280010
Lot NumberA0440680 Certificate TypeCertificate of Analysis Date30/03/2022 Catalog Number388280010
Lot NumberA0434428 Certificate TypeCertificate of Analysis Date21/09/2021 Catalog Number388280010
compliance-icons
Product Identifier
  • L(+)-Tartaric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : (+)-Tartaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L(+)-Tartaric acid, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.