missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine, 99%, Thermo Scientific Chemicals

£21.70 - £186.00

Chemical Identifiers

CAS 20859-02-3
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064218
InChI Key NPDBDJFLKKQMCM-SGAVLPGINA-N
Synonym l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
PubChem CID 164608
IUPAC Name (2S)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)[C@H](N)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13707.03
1 g
£21.70
1g
Please to purchase this item. Need a web account? Register with us today!
11313197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13707.06
5 g
£44.50
5g
Please to purchase this item. Need a web account? Register with us today!
11323197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L13707.14
25 g
£186.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L-tert-Leucine is an essential amino acid and makes up one third of our muscle protein. It can be used as a food additive. It is used in the formation of sterols. It is used in the production of cobalt oxazoline palladacycles complex, which acts as a catalyst in the rearrangement of prochiral N-aryl trifluoroacetimidates to allylic amides yielded in significantly higher enantioselectivities with the (S)-(pR)-diasteroisomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-tert-Leucine is an essential amino acid and makes up one third of our muscle protein. It can be used as a food additive. It is used in the formation of sterols. It is used in the production of cobalt oxazoline palladacycles complex, which acts as a catalyst in the rearrangement of prochiral N-aryl trifluoroacetimidates to allylic amides yielded in significantly higher enantioselectivities with the (S)-(pR)-diasteroisomer.

Solubility
Soluble in water and hydrchloric acid.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

20859-02-3
131.18
NPDBDJFLKKQMCM-SGAVLPGINA-N
164608
CC(C)(C)[C@H](N)C(O)=O
C6H13NO2
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
(2S)-2-amino-3,3-dimethylbutanoic acid

Specifications

>300°C
C6H13NO2
1721824
Soluble in water and hydrchloric acid.
CC(C)(C)[C@H](N)C(O)=O
131.18
131.18
+7° (c=5 in 5N HCl)
20859-02-3
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
NPDBDJFLKKQMCM-SGAVLPGINA-N
(2S)-2-amino-3,3-dimethylbutanoic acid
164608
99%
L-tert-Leucine
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 424-750-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only