missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine methyl ester hydrochloride, 97%

Product Code. 15475928
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15475928

Brand: Thermo Scientific Alfa Aesar H59563.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63038-27-7 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 183°C
CAS 63038-27-7
Molecular Formula C7H16ClNO2
MDL Number MFCD00040604
Beilstein 4879785
Synonym l-tert-leucine methyl ester hydrochloride, h-tle-ome.hcl, s-methyl 2-amino-3,3-dimethylbutanoate hydrochloride, methyl 2s-2-amino-3,3-dimethylbutanoate hydrochloride, s-2-amino-3,3-dimethylbutyric acid methyl ester hydrochloride, methyl 3-methyl-l-valinate hydrochloride, pubchem11076, methyl l-tert-leucinate hydrochloride, l-tert-leucine methyl ester hcl, l-tert-leucine methyl-ester hydrochloride
InChI Key HRTQWUHFSXVRPY-XZNNNFJINA-N
SMILES Cl.COC(=O)[C@@H](N)C(C)(C)C
IUPAC Name methyl (2S)-2-amino-3,3-dimethylbutanoate;hydrochloride
Molecular Weight (g/mol) 181.66
Quantity 1 g
PubChem CID 16217630
Formula Weight 181.66
Optical Rotation +18° (c=1 in Methanol)
Percent Purity 97%
Chemical Name or Material L-tert-Leucine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.