missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Threonine, 98%
£17.29 - £513.56
Chemical Identifiers
CAS | 72-19-5 |
---|---|
Molecular Formula | C4H9NO3 |
Molecular Weight (g/mol) | 119.12 |
MDL Number | MFCD00064270 |
InChI Key | AYFVYJQAPQTCCC-UHFFFAOYNA-N |
Synonym | l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin |
PubChem CID | 6288 |
ChEBI | CHEBI:16857 |
SMILES | CC(O)C(N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10435115
|
Thermo Scientific Chemicals
138930050 |
5 g | Glass Bottle |
£17.29
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10665762
|
Thermo Scientific Chemicals
138930250 |
25 g | Glass Bottle |
£47.44
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001610
|
Thermo Scientific Chemicals
138931000 |
100 g | Glass Bottle |
£139.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10398743
|
Thermo Scientific Chemicals
138935000 |
500 g | Glass Bottle |
£513.56
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72-19-5 | |
119.12 | |
AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
6288 | |
CC(O)C(N)C(O)=O |
C4H9NO3 | |
MFCD00064270 | |
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
CHEBI:16857 |
Specifications
255.0°C | |
Yellow | |
10ppm max. | |
98% | |
C4H9NO3 | |
MFCD00064270 | |
300ppm max. | |
0.1 % max. | |
Solubility in water: 90g/L (20°C). Other solubilities: insoluble in most common organic solvents | |
CC(O)C(N)C(O)=O | |
−28.30 (20.00°C c=6,H2O) | |
6288 | |
119.12 | |
Crystalline Powder |
72-19-5 | |
200ppm max. | |
Authentic | |
0.5 % max. (1 g, 105°C, 3 hrs) | |
CH3CH(OH)CH(NH2)CO2H | |
− 28.30 | |
15, 9534 | |
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
2-amino-3-hydroxybutanoic acid | |
119.12 | |
CHEBI:16857 | |
98% | |
L-Threonine |
RUO – Research Use Only