missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
L-Threonine methyl ester hydrochloride is used as a feed and food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Threonine methyl ester hydrochloride is used as a feed and food additive.
Solubility
Soluble in water.
Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.
L-Threonine methyl ester hydrochloride is used as a feed and food additive.
Solubility
Soluble in water.
Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Beilstein | 3562717 |
Solubility Information | Soluble in water. |
Formula Weight | 169.61 |
Percent Purity | 98% |
Quantity | 25 g |
Sensitivity | Hygroscopic; Moisture sensitive |
Chemical Name or Material | L-Threonine methyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'