missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Specifications
CAS | 72-19-5 |
Molecular Formula | C4H9NO3 |
Packaging | Poly Bottle |
Synonym | l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin |
InChI Key | AYFVYJQAPQTCCC-UHFFFAOYNA-N |
SMILES | CC(O)C(N)C(O)=O |
IUPAC Name | (2S,3R)-2-amino-3-hydroxybutanoic acid |
Molecular Weight (g/mol) | 119.12 |
PubChem CID | 6288 |
ChEBI | CHEBI:16857 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'