missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Thymidine
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Format | Crystalline Powder |
Molecular Weight (g/mol) | 242.23 |
Product Type | L-Thymidine |
Solution Clarity | (1% aq. soln.) Clear colorless solution |
Source | synthetic source |
Content And Storage | Refrigerator +4°C |
Shipping Condition | normal conditions |
Purity | ≥98% by HPLC |
Quantity | 1 g |
Color | White to almost white |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'