missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophyl diphenylborinate, 95%, Thermo Scientific™

Product Code. 15402048
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16655-49-5
Molecular Formula C23H21BN2O2
Molecular Weight (g/mol) 368.243
MDL Number MFCD22125148
InChI Key XBYDPGDPEUHTPB-NRFANRHFSA-N
Synonym l-tryptophyl diphenylborinate, diphenylboranyl 2s-2-amino-3-1h-indol-3-yl propanoate
PubChem CID 99738147
IUPAC Name diphenylboranyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate
SMILES B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CNC4=CC=CC=C43)N

Specifications

Quantity 250 mg
Formula Weight 368.24
Percent Purity 95%
Chemical Name or Material L-Tryptophyl diphenylborinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'