missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophyl diphenylborinate, 95%, Thermo Scientific™
£97.40 - £315.00
Chemical Identifiers
CAS | 16655-49-5 |
---|---|
Molecular Formula | C23H21BN2O2 |
Molecular Weight (g/mol) | 368.243 |
MDL Number | MFCD22125148 |
InChI Key | XBYDPGDPEUHTPB-NRFANRHFSA-N |
Synonym | l-tryptophyl diphenylborinate, diphenylboranyl 2s-2-amino-3-1h-indol-3-yl propanoate |
PubChem CID | 99738147 |
IUPAC Name | diphenylboranyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate |
SMILES | B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CNC4=CC=CC=C43)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16655-49-5 | |
368.243 | |
XBYDPGDPEUHTPB-NRFANRHFSA-N | |
99738147 | |
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CNC4=CC=CC=C43)N |
C23H21BN2O2 | |
MFCD22125148 | |
l-tryptophyl diphenylborinate, diphenylboranyl 2s-2-amino-3-1h-indol-3-yl propanoate | |
diphenylboranyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate |
Specifications
16655-49-5 | |
MFCD22125148 | |
XBYDPGDPEUHTPB-NRFANRHFSA-N | |
diphenylboranyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate | |
99738147 | |
95% |
C23H21BN2O2 | |
l-tryptophyl diphenylborinate, diphenylboranyl 2s-2-amino-3-1h-indol-3-yl propanoate | |
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CNC4=CC=CC=C43)N | |
368.243 | |
368.24 | |
L-Tryptophyl diphenylborinate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures