missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent Product Code.: 15405729

Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent

Product Code. 15405729
10 g
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15405729

Brand: Thermo Scientific Chemicals J63511.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, lShow MoreShow Less
PubChem CID 6057
ChEBI CHEBI:17895
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point >300°C
Color Brown to White
Beilstein 392441
Merck Index 14,9839
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Quantity 10 g
Formula Weight 181.19
Optical Rotation −11° (c=4 in 1N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Tyrosine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU29J008 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ63511.09,J63511.36,J63511.22,J63511.A1
Lot NumberU08I019 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog NumberJ63511.09,J63511.36,J63511.22,J63511.A1
Lot NumberU28D036 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ63511.09,J63511.36,J63511.22,J63511.A1
Lot NumberY15H093 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ63511.09,J63511.36,J63511.22,J63511.A1
Lot NumberN11H026 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63511.09,J63511.36,J63511.22,J63511.A1
compliance-icons
Product Identifier
  • L-Tyrosine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : L-Tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.