missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine disodium salt, 98%

Product Code. 10306734 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10306734

Brand: Thermo Scientific Acros 426411000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69847-45-6 | C9H9NNa2O3 | 225.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69847-45-6
Molecular Formula C9H9NNa2O3
Molecular Weight (g/mol) 225.16
MDL Number MFCD00037203
InChI Key ASIYFCYUCMQNGK-RIYJSUBBNA-L
Synonym sodium s-2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 66602062
SMILES [Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Melting Point >300.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula NaOC6H4CH2CH(NH2)CO2Na
Specific Rotation − 12.90
IUPAC Name disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate
Quantity 100 g
Formula Weight 225.15
Percent Purity 98%
Chemical Name or Material L-Tyrosine disodium salt
compliance-icons
Product Identifier
  • L-Tyrosine disodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.