missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine disodium salt dihydrate, Thermo Scientific Chemicals

Catalog Number p-7042223
Click to view available options
Quantity:
100 g
500 g
1 kg
122666-87-9
C9H13NNa2O5
261.19
MFCD03840558
WFTCFVUQORELJZ-CMIWSEPMNA-L
tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate
71587456
O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O
122666-87-9
C9H13NNa2O5
261.19
MFCD03840558
WFTCFVUQORELJZ-CMIWSEPMNA-L
tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate
71587456
O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color Brown to White
Solubility Information Soluble in water.
IUPAC Name 2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium
Formula Weight 261.18 (225.15 Anhydrous)
Percent Purity 99%
Quantity 100 g
Optical Rotation −13° (c=4 in Water)
Sensitivity Very Hygroscopic
Physical Form Powder
Chemical Name or Material L-Tyrosine disodium salt dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'