missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.
Solubility
Soluble in water (3.685e+005 mg/L @ 25°C (est.)).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.
Solubility
Soluble in water (3.685e+005 mg/L @ 25°C (est.)).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 166°C to 168°C |
Beilstein | 4725904 |
Solubility Information | Soluble in water (3.685e+005mg/L @ 25°C (est.)). |
Formula Weight | 245.71 |
Percent Purity | 99% |
Quantity | 25 g |
Optical Rotation | −6.8° (c=2 in Water) |
Chemical Name or Material | L-Tyrosine ethyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'