missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine methyl ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 134°C to 136°C |
Beilstein | 2372626 |
Solubility Information | Soluble in water. |
Formula Weight | 195.22 |
Percent Purity | 98% |
Quantity | 25 g |
Chemical Name or Material | L-Tyrosine methyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'