missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valinamide hydrochloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity
Solubility
Soluble in methanol (50 mg/ ml-clear, colorless solution).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity
Solubility
Soluble in methanol (50 mg/ ml-clear, colorless solution).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 266°C to 270°C |
Beilstein | 4198583 |
Solubility Information | Soluble in methanol (50mg/ ml-clear,colorless solution). |
Formula Weight | 152.62 |
Percent Purity | 95% |
Quantity | 5 g |
Physical Form | Solid |
Chemical Name or Material | L-Valinamide hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'