missing translation for 'onlineSavingsMsg'
Learn More

L-Valine, Cell Culture Reagent, Thermo Scientific Chemicals

Product Code. 15425249
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Color White
Beilstein 1721136
Merck Index 14,9909
Formula Weight 117.15
Grade Cell Culture Reagent
Quantity 1 kg
Odor Odorless
Physical Form Powder
Chemical Name or Material L-Valine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'