Learn More
OHAUS™ Lab Lift
Shop All Ohaus Products
Click to view available options
Size:
102 x 102 (L x W) Platform
152 x 152 (L x W) Platform
203 x 203 (L x W) Platform
254 x 254 (L x W) Platform
305 x 305 (L x W) Platform
406 x 406 (L x W) Platform
76 x 76 (L x W) Platform
Material:
Aluminum
Stainless Steel
Unit Size:
Each
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Specifications
| Type | Lab Lift |
| Material | Aluminum |
| Description | Clamp, Support, Lab-Lift, CLR-LBLFTA015 |
| Height (Metric) | 76 to 248 mm |
| Size | 152 x 152 (L x W) Platform |