Learn More
OHAUS™ Lab Lift
Product Code. 15752419 Shop All Ohaus Products
Click to view available options
Size:
102 x 102 (L x W) Platform
152 x 152 (L x W) Platform
203 x 203 (L x W) Platform
254 x 254 (L x W) Platform
305 x 305 (L x W) Platform
406 x 406 (L x W) Platform
76 x 76 (L x W) Platform
Material:
Aluminum
Stainless Steel
missing translation for 'unitSize'
Each
Product Code. 15752419
missing translation for 'mfr': OHAUS™ 30400003
Only null left
Add to basket
Add to basket
OHAUS™ Lab Lift provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Type Lab Lift
Material Stainless Steel
Description Clamp, Support, Lab-Lift, CLR-LBLFTS020
Height (Metric) 76 to 248 mm
Size 203 x 203 (L x W) Platform