missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle Lab Packs with Screw-On Lid
Eagle Lab Packs are ideal for safe storage, handling and transportation of hazardous materials, spill containment and clean-up of hazardous materials, original shipping container, bio waste disposal and incineration.
£93.90 - £222.00
Specifications
Color | Yellow |
---|---|
For Use With (Application) | Safe Storage, Handling and Transportation of Hazardous Materials |
Material | Polyethylene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17752308
|
Eagle
EDR16500YL |
75.7 L |
£93.90
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17742308
|
Eagle
EDR1600SYL |
113 L |
£106.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17762308
|
Eagle
EDR16570YL |
217 L |
£222.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Eagle Lab Pack Drums are constructed of Yellow blow-molded, high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, weather and chemical resistant, and screw-on lid
- Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
- Lab Packs/Open Head Drums- for use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS
Specifications
Yellow | |
Safe Storage, Handling and Transportation of Hazardous Materials | |
Polyethylene |