missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine Lab Sink with Drain
Lab Sink with Drain for integration in fume cupboards or on worktops for easy disposal of liquid solvent waste in large quantities.
Brand: SymLine 118020
524.51 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Made of dissipative PE-HD-EL
- Base with integrated slope for complete emptying
- Drain with removable sieve
Specifications
Black | |
Laboratory Sink | |
400 x 300 x 190 mm |
HDPE | |
G 1.5 in. OD Thread |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction