Learn More
Berkshire Lab-Tips™ General Purpose Swab
The LTA03163 Swab is an excellent general-purpose swab for cleaning small and confined areas.
Brand: Berkshire LTAO3163.10
This item is not returnable.
View return policy
Description
- Compressed, 100 ppi polyurethane foam
- Thermally bonded head
- Polyurethane foam over rigid core
- Low particles, fibers, ions and extractables
- Good chemical resistance
- Soft and non-abrasive with no contaminating adhesives
- Good sorbency
Specifications
3 in. | |
Not Autoclavable | |
Thermal | |
Rigid |
Cleanroom Swab | |
Non-sterile | |
Plastic | |
Foam |