Learn More
Berkshire™ Lab-Tips™ Low TOC Swab Large
Berkshire Lab-Tips™ Low TOC Swab Large knitted polyester swab features a unique semi-flexible internal paddle for greater surface coverage.
Brand: Berkshire™ LTP125TOC.25
Additional Details : Weight : 0.25000kg
Description
- Specifically designed for Total Organic Carbon analysis and TOC tests in low-TOC environment
- Critically low particles, fibers, ions and extractables
- Excellent chemical resistance
- High abrasion resistance while soft and non-abrasive
- No contaminating adhesives and good sorbency
Specifications
20 swabs/pack; 25 packs/case | |
TOC Swab | |
Polyester |
3.94 in. | |
100mm | |
Large |