Learn More
Berkshire Lab-Tips™ Pointed Head Swab
The LTO70P Swab with pointed head is an excellent swab for precision cleaning applications.
Brand: Berkshire LT070P20
This item is not returnable.
View return policy
Description
- Cleanroom laundered polyurethane foam over rigid tip
- 100 ppi open cell foam
- Polypropylene handle with thermally bonded head
- Laundered and packaged in Class 10 {ISO Class 4} cleanroom
- Low particles, fibers, ions and extractables
- Good chemical resistance
- Soft and non-abrasive with no contaminating adhesives
- Free from Silicone, amides and DOP
- Ridged surface for improved cleaning ability
Specifications
2.75 in. | |
Not Autoclavable | |
Plastic | |
Foam |
Cleanroom Swab | |
Thermal | |
Pointed |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Berkshire Lab-Tips™ Pointed Head Swab > Berkshire Lab-Tips™ Pointed Head Swab
Spot an opportunity for improvement?Share a Content Correction