missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Calcitonin Rabbit Monoclonal Antibody
Identify a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors using Epredia™ Calcitonin Rabbit Monoclonal Antibody.
Brand: Epredia™ RM-9117-S
This item is not returnable.
View return policy
Description
Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.Host Species: Rabbit
Clone: SP17
Speices Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.
Epitope: 1-32aa
Immunogen: Synthetic human calcitonin 1-32 amino acid peptide
Positive Control: Thyroid or medullary CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Calcitonin | |
Monoclonal | |
Unconjugated | |
Synthetic human calcitonin 1-32 amino acid peptide | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
SP17 | |
Rabbit | |
1 mL | |
Steroids Hormones and Growth Factors | |
Canine, Equine, Human, Mouse, Non-human Primate, Porcine, Rat, Sheep |