missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calcitonin Rabbit Monoclonal Antibody

Identify a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors using Epredia™ Calcitonin Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-9117-S0

 View more versions of this product

Product Code. 12683747

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.

Host Species: Rabbit

Clone: SP17

Speices Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Epitope: 1-32aa

Immunogen: Synthetic human calcitonin 1-32 amino acid peptide

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Calcitonin
Monoclonal
Unconjugated
Synthetic human calcitonin 1-32 amino acid peptide
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
SP17
Rabbit
100 μL
Steroids Hormones and Growth Factors
Canine, Equine, Human, Mouse, Non-human Primate, Porcine, Rat, Sheep
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers