missing translation for 'onlineSavingsMsg'
Learn More

CD137 (4-1BB) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ CD137 (4-1BB) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-621-P

 View more versions of this product

Product Code. 12621817

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

4-1BB (also known as CD137) is an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. It exists as both a monomer and a dimer on the surface of activated T cells. 4-1BB is structurally related to the members of NGFR/TNFR superfamily which are characterized by the presence of three-six patterns of a cysteine-rich motif in their extracellular domains. Other members of this family include low affinity NGFR, two receptors for TNF (TNFR-I & TNFR-II), CD30, CD40, OX40, Fas, and CD27. These molecules are involved in cell growth, survival, and death processes. The cytoplasmic domain of 4-1BB include two runs of acidic amino acids, a potential p56lck binding site, five consecutive glycines at the C-terminus, and four potential phosphorylation sites: one tyrosine, two threonine, and one serine.

Host Species: Mouse

Clone: BBK-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Ectodomain

Immunogen: Ectodomain of human 4-1BB recombinant protein

Molecular Weight: 30kDa

Positive Control: Spleen, thymus or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD137 Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Apoptosis
Human
IgG1 κ
Flow Cytometry, Immunofluorescence
BBK-2
Unconjugated
Ectodomain of human 4-1BB recombinant protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers