missing translation for 'onlineSavingsMsg'
Learn More

CD29 (Integrin β-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments with Epredia™ CD29 (Integrin β-1) Ab-1.

Brand:  Epredia MS-596-P0

 View more versions of this product

Product Code. 12641707

  • £119.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD29 (integrin Beta-1/gpIIa) is a cell adhesion molecule appearing on platelets, as the common Beta subunit of the very late activation antigen (VLA), and as a component of various protein complexes binding to extracellular matrix proteins. CD29 is expressed at the cell surface exclusively as part of a heterodimer, in association with one of at least nine different integrin alpha subunits (alpha1, alpha2, alpha3, alpha4, alpha5, alpha6, alpha7, alpha8, and alphav). With the exception of red blood cells and possible weak expression on granulocytes, CD29 is expressed on nearly all cell and tissue types.

Host Species: Mouse

Clone: 4B7R

Isotype: IgG1/κ

Species Reactivity: Human and Pig. Others not tested.

Epitope: Not determined

Immunogen: Human melanoma V+B2 cell line

Molecular Weight: 110kDa (nonreducing) and 130 (reducing)

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD29 (Integrin beta-1) Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cell Adhesion
Human, Porcine
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
4B7R
Unconjugated
Human melanoma VplusB2 cell line
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers