Learn More
Epredia™ Lab Vision™ CD3 (Early T-Cell Marker) Rabbit Monoclonal Antibody,
Stain normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues with Epredia™ CD3 (Early T-Cell Marker), Rabbit Monoclonal Antibody.
Brand: Epredia™ RM-9107-S
1339.50 GBP valid until 2024-12-31
Use promo code "21882" to get your promotional price.
Code : LD
Additional Details : Weight : 0.00250kg
Description
This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.Host Species: Rabbit
Clone: SP7
Isotype: IgG
Species Reactivity: Human, Baboon, Monkey, Horse, Dog and Cat. Others not tested.
Epitope: 156-168aa
Immunogen: A synthetic 13-mer peptide corresponding to aa 156-168 of the ε-chain of human CD3 protein
Positive Control: Jurkat cells, Tonsil
Cellular Localization: Membrane
Clone SP7 is excellent for staining of formal/paraffin.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD3 | |
Monoclonal | |
Unconjugated | |
A synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon-chain of human CD3 protein | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
SP7 | |
Rabbit | |
1 mL | |
Cancer and Tumor Biology | |
Canine, Equine, Feline, Human, Non-human Primate | |
IgG |