Learn More
Epredia™ Lab Vision™ CD5 Ab-1, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD5 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-393-R7
272.80 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
CD5, a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphoma, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.
Host Species: Mouse
Clone: 4C7
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: External domain
Immunogen: Recombinant protein corresponding to the external domain of the CD5 molecule
Molecular Weight: 67kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Specifications
CD5 Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to the external domain of the CD5 molecule | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
4C7 | |
Mouse | |
7 mL | |
Hematopoietic | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.