Learn More
Epredia™ Lab Vision™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody.
£137.00 - £512.00
Specifications
Antigen | CD71/Transferrin Receptor Ab-3 |
---|---|
Clone | 10F11 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12678936
|
Epredia™
MS-1096-S0 |
Concentrated | 100 μL | N/A | |||||
12668936
|
Epredia™
MS-1096-S1 |
Concentrated | 500 μL |
£342.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12658936
|
Epredia™
MS-1096-S |
Concentrated | 1 mL |
£512.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12648936
|
Epredia™
MS-1096-R7 |
Prediluted | 7 mL |
£488.00
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cell surface transferrin receptor (CD71) is a disulfide-bonded homodimeric glycoprotein. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. Transferrin receptor (CD71) has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.Host Species: Mouse
Clone: 10F11
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Molecular Weight: ∽90-95 kDa
Positive Control: Placenta
Cellular Localization: Granular cytoplasmic and membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
General References:1. Habeshaw J A, et al. Lancet. 1:498-500 (1983)
2. Lioyd J M, et al. Journal of Clinical Pathology. 37: 131-135 (1984).
Specifications
CD71/Transferrin Receptor Ab-3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
IgG2b |
10F11 | |
Monoclonal | |
Mouse | |
Human | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.