missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CDX2 Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CDX2, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-2116-S0

 View more versions of this product

Product Code. 12653657

  • £332.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The caudal-related homeodomain protein2 (CDX2) is a transcription factor shown to play a role in the development of small and large intestine in mammals and in the differentiation of intestinal epithelial cells. It has been shown that CDX2 protein detection by IHC coorelates with RNA transcript levels. In studies using 745 cancers from many anatomic sites, colonic adenocarcinomas demonstrated strong staining in 90% of the cases, while adenocarcinomas of the stomach, esophagus and ovary showed extensive staining in only 30% of the cases. Stains of CDX2 and Villin are useful markers in differntial diagnosis of bladder adenocarcinoma and secondary colorectal adenocarcinoma.

Host Species: Rabbit

Clone: EPR2764Y

Species Reactivity: Human. Others not tested.

Ig Isotype: Rabbit IgG

Epitope: not determined

Immunogen: A synthetic peptide corresponding to residues near the N-terminus of human CDX2

Molecular Weight: 40 kDa

Positive Control: Colon carcinomas

Cellular Localization: Nuclear with occasional cytoplasmic staining on colon carcinoma

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

anti-CDX2
Monoclonal
Unconjugated
A synthetic peptide corresponding to residues near the N-terminus of human CDX2
Primary
Supernatant
Immunohistochemistry
EPR2764Y
Rabbit
100 μL
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.