Learn More
Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X)
Improve the reactivity of many antibodies in formalin-fixed tissues with the Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X).
Brand:  Thermo Scientific™ TA-050-CBX
Product Code. 12633947
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Improve the reactivity of many antibodies in formalin-fixed tissues with the Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X)
  • 1M Citrate buffer, pH 6.0
  • 100X stock solution
  • Diluted 100-fold with distilled water before use
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives. For periods longer than 3 months, store at 2-8°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
100X
50 mL
Room temperature
Citrate Buffer
6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X) > 50mL