missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X)
Improve the reactivity of many antibodies in formalin-fixed tissues with the Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X).
£99.50 - £222.00
Specifications
Content And Storage | Room temperature |
---|---|
Dilution | 10X |
Product Type | Citrate Buffer |
For Use With (Application) | Immunohistochemistry |
pH | 6 |
Description
- Citrate buffer, pH 6.0
- 10X stock solution
- Diluted 10-fold with distilled water before use
- Store at room temperature.
- Stable for up to 18 months.
- Contains no preservatives.
- For periods longer than 3 months, store at 2-8°C.
Specifications
Room temperature | |
Citrate Buffer | |
6 |
10X | |
Immunohistochemistry |
For Reserach Use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X)