Learn More
Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X)
Improve the reactivity of many antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X).
Brand: Epredia™ AP-9004-500
This item is not returnable.
View return policy
Description
- 10mM EDTA buffer, pH 8.0
- Dilute 10-fold with distilled water before use
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives. For periods longer than 3 months, store at 2-8°C
Specifications
Room temperature | |
EDTA | |
Immunohistochemistry |
10X | |
500mL | |
8 |
Product Content Correction
Product Title
Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X) > 500mL
Spot an opportunity for improvement?Share a Content Correction