missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ HIER Buffer L (100X)
Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Epredia™ Lab Vision™ HIER Buffer L (100X).
Brand: Epredia™ TA-135-HBL
This item is not returnable.
View return policy
Description
- For in vitro diagnostic use
- Storage Conditions: Store at room temperature

Specifications
Room temperature | |
HIER | |
Immunohistochemistry | |
IVD |
100X | |
9 x 15 mL | |
6 to 7 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Lab Vision™ HIER Buffer L (100X) > 9 x 15mL
Spot an opportunity for improvement?Share a Content Correction