Learn More
Epredia™ Lab Vision™ HIER Buffer M (100X)
Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Epredia™ Lab Vision™ HIER Buffer M (100X)
Brand: Epredia™ TA-135-HBM
Description
- For in vitro diagnostic use
- Store at room temperature

Specifications
Room temperature | |
HIER | |
Immunohistochemistry | |
IVD |
100X | |
9 x 15 mL | |
6 to 7 |
Safety and Handling
- X9 HIER BUFFER M, 9 BT/CS, (100X CONCENTRATE), 100X stock solution, must be diluted 100 fold
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : EDTA, Tetrasodium EDTA, 2-Methyl-3-isothiazolone
Your input is important to us. Please complete this form to provide feedback related to the content on this product.