missing translation for 'onlineSavingsMsg'
Learn More

Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 Mouse Monoclonal Antibody, Epredia™

Deliver accurate, reproducible results with the Epredia™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-145-P0

 View more versions of this product

Product Code. 12659866

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.

Host Species: Mouse

Clone: 45M1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog and Chicken. Does not react with cow. Others not known.

Epitope: Destroyed by β-mercaptoethanol and proteases but not by periodate treatment

Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient

Molecular Weight: >1000kDa

Positive Control: Stomach

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Chicken, Feline, Hedgehog, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat
IgG1 κ
Immunohistochemistry (Paraffin)
45M1
Unconjugated
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers