missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)

Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).

£36.25 - £201.00

Specifications

Product Type PBS and Tween
For Use With (Application) Anatomical Pathology
Concentration 20X
Content And Storage Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12673967
View Documents
Epredia™
TA-125-PT
125 mL
£36.25
125mL
Please to purchase this item. Need a web account? Register with us today!
12364443
View Documents
Epredia™
TA-999-PT
1000 mL
£201.00
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For in vitro diagnostic use
  • Added to promote more effective washings, resulting in decreased nonspecific background staining
  • Aids reagent spreading on slides

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.
Specifications

Specifications

PBS and Tween
Anatomical Pathology
20X
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For in vitro diagnostic use