missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval

Improve the immunoreactivity of certain antigens in formaldehyde-fixed tissues with Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval.

Brand:  Epredia™ AP-9006-005

 View more versions of this product

Product Code. 12618296

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Storage Conditions: Store at 2°-8°C. Stable for 18 months
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C.
Protease
Immunohistochemistry
1 mg/mL
5mL
7.4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval > Reconstitute to 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For in vitro diagnostic use