missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval
Improve the immunoreactivity of certain antigens in formaldehyde-fixed tissues with Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval.
£87.43
Specifications
Content And Storage | 2° to 8°C. |
---|---|
Concentration | 1 mg/mL |
Product Type | Protease |
For Use With (Application) | Immunohistochemistry |
pH | 7.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12618296
|
Epredia™
AP-9006-005 |
5mL | N/A | ||||||
Description
Storage Conditions: Store at 2°-8°C. Stable for 18 monthsSpecifications
2° to 8°C. | |
Protease | |
7.4 |
1 mg/mL | |
Immunohistochemistry |
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval