missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions (100X), Citrate buffer, pH 6

Perform Heat Induced Antigen Retrieval (HIER) on formalin fixed paraffin-embedded tissue sections with Epredia™ Lab Vision™ PT Module™ Epitope Retrieval Solutions.

£324.17 - £609.76

Specifications

Content And Storage Room temperature
Dilution 100X
Product Type HIER
For Use With (Application) Immunohistochemistry
View More Specs

Products 5
Product Code Brand pH Quantity Price Quantity & Availability  
Product Code Brand pH Quantity Price Quantity & Availability  
12633967
View Documents
Epredia™
TA-125-PM1X
6 125mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12643967
View Documents
Epredia™
TA-125-PM2X
8 125mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12663967
View Documents
Epredia™
TA-125-PM4X
9 125mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12633987
View Documents
Epredia™
TA-250-PM1X
6 250mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12643987
View Documents
Epredia™
TA-250-PM2X
8 250mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • 100X stock solution
  • Dilute 100-fold with distilled water before use
  • Designed for use with Pretreatment Module™
  • Available in Citrate, EDTA or Tris-HCl at various pH levels

Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives; for periods longer than three months, store at 2°-8°C.

Specifications

Specifications

Room temperature
100X
HIER
Immunohistochemistry
Videos
SDS
Documents

Documents

Product Certifications