missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Ready-To-Use Streptavidin Peroxidase
Epredia™ Lab Vision Steptravidin Peroxidase (Ready-to-Use) reagent is intended for use in a labeled streptavidin-biotin immunoenzymatic antigen detection system.
Brand: Epredia™ TS-060-HR
Code : LD
Additional Details : Weight : 0.11340kg
Description
This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate-chromogen.Specifications
Streptavidin Peroxidase (Ready-To-Use) | |
2deg;C to 8°C. Each component is stable for 18 months. | |
Streptavidin Peroxidase |
60 mL | |
Lab Vision™ |