missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lab Vision™ Tris Buffered Saline and Tween 20 (20X), Epredia™
Take advantage of a wash buffer for immunolabeling techniques with the Utilize Epredia™ Lab Vision™ Tris-Buffered Saline and Tween™ 20 (20X).
£28.55 - £366.00
Specifications
Product Type | TBS with Tween (TBST) |
---|---|
For Use With (Application) | Anatomical Pathology |
Concentration | 20X |
Content And Storage | Room Temperature |
Description
- Tween 20 is added to promote more effective washings, resulting in decreased nonspecific background staining
- Tween 20 aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week
Specifications
TBS with Tween (TBST) | |
Anatomical Pathology | |
20X | |
Room Temperature |
For in vitro diagnostic use