missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X)
Improve the reactivity of certain antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X).
£51.00 - £179.00
Specifications
Content And Storage | 2° to 8°C. |
---|---|
Dilution | 10X |
Product Type | HIER |
For Use With (Application) | Immunohistochemistry |
pH | 10 |
Description
- 100X stock solution
- 100mM Tris-HCl buffer, pH 10.0
- Dilute 10-fold with distilled water before use
Storage Conditions: Store at 2-8°C. Stable for up to 18 months. Contains no preservatives.
Specifications
2° to 8°C. | |
HIER | |
10 |
10X | |
Immunohistochemistry |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Tris-HCl Buffer
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H331-Toxic if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P321-Specific treatment (see on this label).
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Tris (hydroxymethyl)aminomethane, Hydrogen chloride
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X)