Learn More
Epredia™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer (Ready-To-Use)
A one-step polymer system providing increased sensitivity, time savings and detection simplicity.
£955.00 - £1910.00
Specifications
Chromagen | None provided |
---|---|
Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
Storage Requirements | Store at 2° to 8°C. Each component is stable for 18 months. |
Enzyme | Peroxidase |
Substrate | None provided |
Description
UltraVision ONE is a robust ONE-step polymer system that provides increased sensitivity, time savings and detection simplicity. The UltraVision ONE HRP polymer is an innovative, patented technology. It consists of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. UltraVision ONE HRP polymer allows the use of less antibody to obtain better signal-to-noise ratios. This system is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods. For optimal interpretation of results, appropriate positive and negative controls must be included.Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: None provided
Includes:
TL-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE HRP Polymer (TL-060-PHJ)
TL-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE HRP Polymer (TL-125-PHJ)
Note:
UltraVision ONE HRP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to horseradish peroxidase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
None provided | |
Store at 2° to 8°C. Each component is stable for 18 months. | |
None provided |
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) | |
Peroxidase |