missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Vimentin Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-129-P1ABX
This item is not returnable.
View return policy
Description
Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.Host Species: Mouse
Clone: V9
Isotype: IgG1/κ
Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.
Epitope: Not determined
Immunogen: Purified vimentin from pig eye lens
Molecular Weight: 57-60kDa
Positive Control: Sarcomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Vimentin Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cytoskeletal Protein | |
Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
V9 | |
Unconjugated | |
Purified vimentin from pig eye lens | |
IVD | |
Primary | |
Concentrated |