missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Ab-2 Mouse Monoclonal Antibody, Epredia™

Epredia™ Vimentin Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-129-B1

 View more versions of this product

Product Code. 12689506

  • £379.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Mouse

Clone: V9

Isotype: IgG1/κ

Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.

Epitope: Not determined

Immunogen: Purified vimentin from pig eye lens

Molecular Weight: 57-60kDa

Positive Control: Sarcomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Vimentin Ab-2
Monoclonal
200μg/mL
Mouse
500 μL
Cytoskeletal Protein
Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat
IgG2a κ
Immunohistochemistry (Paraffin)
V9
Biotin
Purified vimentin from pig eye lens
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers