missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Vinculin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ Vinculin Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1209-P0

 View more versions of this product

Product Code. 12649266

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Vinculin is an attachment protein involved in the indirect binding of intracellular actin filaments to extracellular fibronectin. Vinculin binds to actin capping protein and α actinin, which in turn binds to actin. The binding of fibronectin is also indirect where vinculin binds to talin, which in turn binds to the intracellular domain of the fibronectin receptor which itself is the ligand for fibronectin. Vinculin is widely distributed in tissue and is expressed where smooth muscle actin and fibroblasts attach to the extracellular matrix.

Host Species: Mouse

Clone: VLN01

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified vinculin

Molecular Weight: 130kDa (vinculin) and 150kDa (meta-vinculin)

Positive Control: HeLa cells, skin

Cellular Localization: Cell membrane and cytoplasmic Ab-1 is useful for studies of vinculin in platelet activation.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Vinculin Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cytoskeletal Protein
Human
IgG
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
VLN01
Unconjugated
Semi-purified vinculin
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions