missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ CentriVap™ Concentration Systems: -50°C Cold Trap

Cold trap for use with Labconco CentriVap Benchtop Concentrator

Brand:  Labconco™ 7811030

 View more versions of this product

Product Code. 10368762

  • £4930.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Protect vacuum pump by trapping moisture, vapors and corrosive fumes as they evaporate from samples
  • Environment-friendly, CFC-free refrigerant
  • Stainless-steel condenser collects 4L of liquid, accepts optional glass insert (16-315-61) for corrosives
  • Corrosion-resistant drain speeds cleanup
  • READY light illuminates at -40°C, indicating concentration conditions are right

Order Info

Available in 115V and 230V models.

Compliance

ETL, ETL-C, and CE listed

Specifications

Specifications

CE, ETL, ETL-C
Aqueous concentrator system
Stainless Steel, Acrylic Lid
NEMA 6-15
14.5 in.
13.8 in.
24.2 in.
80 lb.
230 V 50 Hz
2.4 A
-50°C
1/3hp Compressor, 4L Stainless Steel collector, Acrylic lid with two quick-disconnect fittings, Stainless Steel drain port with Nylon valve, grounding clip and on/off power switch
-40°C
Benchtop
36.8 cm
35 cm
61.4 cm
36 kg
230 V
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers